Learn More
Selleck Chemical LLC Nocodazole 10mg 31430-18-9 Oncodazole
SDP
Supplier:  Selleck Chemical LLC S277510MG
Encompass_Preferred
Nocodazole (Oncodazole) is a rapidly-reversible inhibitor of microtubule polymerization, also inhibits Abl, Abl(E255K) and Abl(T315I) with IC50 of 0.21 ?M, 0.53 ?M and 0.64 ?M in cell-free assays, respectively. Nocodazole induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-7956