Learn More
Apexbio Technology LLC Nocodazole 31430-18-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061477
Encompass_Preferred
Nocodazole (CAS 31430-18-9) is an anti-mitotic small molecule commonly utilized in biomedical research as a potent and reversible inhibitor of microtubule polymerization Its mechanism involves direct binding to -tubulin disrupting microtubule assembly and stability It exhibits inhibitory effects on various kinases associated with oncogenesis including Abl c-Kit BRAF and MEK and induces apoptosis in cancer cells In vitro exposure at high concentrations rapidly depolymerizes microtubules while lower concentrations primarily affect their dynamic instability Nocodazole serves as a valuable tool in studies probing microtubule dynamics intracellular trafficking cell cycle regulation and antitumor therapeutic evaluations
Catalog No. 50-000-61477 Supplier Apexbio Technology LLC Supplier No. A84875.1
Nocodazole (CAS 31430-18-9) is an anti-mitotic small molecule commonly utilized in biomedical research as a potent and reversible inhibitor of microtubule polymerization Its mechanism involves direct binding to -tubulin disrupting microtubule assembly and stability It exhibits inhibitory effects on various kinases associated with oncogenesis including Abl c-Kit BRAF and MEK and induces apoptosis in cancer cells In vitro exposure at high concentrations rapidly depolymerizes microtubules while lower concentrations primarily affect their dynamic instability Nocodazole serves as a valuable tool in studies probing microtubule dynamics intracellular trafficking cell cycle regulation and antitumor therapeutic evaluations