Learn More
AdipoGen Nocodazole
SDP
Supplier:  AdipoGen AGCR10019M050
Encompass_Preferred
Chemical. CAS 31430-18-9. Formula C14H11N3O3S. MW 301.3. Microtubule inhibitor. Antitumor compound. Mitosis inhibitor. Arrests the cell cycle at G2/M phase. Promotes tubulin depolymerization. Induces fragmentation of the Golgi complex. Inhibits the T cell antigen receptor. Stimulates the intrinsic GTPase activity of tubulin. Activates the JNK/SAPK signaling pathway. Apoptosis inducer. Autophagy inhibition through prevention of autophagosome-lysosome fusion.
Catalog No. 50-256-2394