Learn More
Selleck Chemical LLC Nocodazole S2775-10mM/1mL
SDP
Catalog No. 503108135
Encompass_Preferred
Nocodazole is a rapidly-reversible inhibitor of microtubule polymerization also inhibits Abl Abl(E255K) and Abl(T315I) with IC50 of 0 21 M 0 53 M and 0 64 M in cell-free assays respectively Nocodazole induces apoptosis
Catalog No. 50-310-8135 Supplier Selleck Chemical LLC Supplier No. S277510MM/1ML
 Restricted Item
Add to Cart
Add to Cart
Nocodazole is a rapidly-reversible inhibitor of microtubule polymerization also inhibits Abl Abl(E255K) and Abl(T315I) with IC50 of 0 21 M 0 53 M and 0 64 M in cell-free assays respectively Nocodazole induces apoptosis