Learn More
Selleck Chemical LLC Nocodazole S2775-50mg
SDP
Catalog No. 503112290
Encompass_Preferred
Nocodazole is a rapidly-reversible inhibitor of microtubule polymerization also inhibits Abl Abl(E255K) and Abl(T315I) with IC50 of 0 21 M 0 53 M and 0 64 M in cell-free assays respectively Nocodazole induces apoptosis
Catalog No. 50-311-2290 Supplier Selleck Chemical LLC Supplier No. S277550MG
Nocodazole is a rapidly-reversible inhibitor of microtubule polymerization also inhibits Abl Abl(E255K) and Abl(T315I) with IC50 of 0 21 M 0 53 M and 0 64 M in cell-free assays respectively Nocodazole induces apoptosis