Learn More
Cayman Chemical Nocodazole, 31430-18-9, 5 mg
SDP
Supplier:  Cayman Chemical 138575
Encompass_Preferred
Molecular Formula C14H11N3O3S, Purity ≥98%, Formula Weight 301.3, Synonyms: NSC 238159, Oncodazole, R 17934; Reversibly binds tubulin and alters GTP hydrolysis as well as microtubule dynamics; nanomolar concentrations reduce both microtubule elongation and shortening velocities, resulting in arrest of cycling in a G2/M phase; also used to induce the formation of Golgi ministacks in eukaryotic cells.
Catalog No. NC1084348