Learn More
Enzo Life Sciences Nocodazole (50 mg). CAS: 31430-18-9
SDP
Catalog No. 502012204
Encompass_Preferred
Synthetic reversible microtubule inhibitor. Alternative name: Methyl[5-(2-thienylcarbonyl)-1H-benzimidazol-2-yl]carbamate, R 17934. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (10mg/ml) or dimethyl formamide; almost insoluble in water. Long Term Storage: Ambient.
Catalog No. 50-201-2204 Supplier Enzo Life Sciences Supplier No. BMLT1010050
Synthetic reversible microtubule inhibitor. Alternative name: Methyl[5-(2-thienylcarbonyl)-1H-benzimidazol-2-yl]carbamate, R 17934. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (10mg/ml) or dimethyl formamide; almost insoluble in water. Long Term Storage: Ambient.