Learn More
Medchemexpress LLC HY-13520 50mg , Nocodazole CAS:31430-18-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1352050MG
Encompass_Preferred
HY-13520 50mg Nocodazole CAS: 31430-18-9 Nocodazole (Oncodazole) is a rapidly-reversible inhibitor of microtubule. Nocodazole binds to β-tubulin and disrupts microtubule assembly/disassembly dynamics, which prevents mitosis and induces apoptosis in tumor cells. Nocodazole inhibits Bcr-Abl, and activates CRISPR/Cas9. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0766