Learn More
Abcam Nocodazole, Microtubule inhibitor, 50MG
Catalog No. 502584580
Encompass_Preferred
MW 301.32 Da, Purity >98%. Microtubule inhibitor. Rapidly depolymerises microtubules in vivo and inhibits tubulin polymerisation in vitro. Inhibits mitosis. Also reduces stem cell pluripotency factors Oct 4 and Nanog.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4580 Supplier Abcam Supplier No. AB12063050MG
MW 301.32 Da, Purity >98%. Microtubule inhibitor. Rapidly depolymerises microtubules in vivo and inhibits tubulin polymerisation in vitro. Inhibits mitosis. Also reduces stem cell pluripotency factors Oct 4 and Nanog.
The product is subject to the following: Abcam Restricted Use Statement