Learn More
STEMCELL Technologies Nocodazole, Size: 50 mg
SDP
Supplier:  STEMCELL Technologies 74074
Encompass
Nocodazole is an anti-mitotic agent that reversibly interferes with the polymerization of microtubules (De Brabander et al.). Nocodazole binds to beta-tubulin and disrupts microtubule assembly/disassembly dynamics, impairing formation of the metaphase spindles during the cell division cycle. This prevents mitosis by inducing a G2/M-phase arrest and induces apoptosis in tumor cells (Jordan et al.).GENOME EDITING· Synchronizes human pluripotent stem cells (hPSCs) in the G2/M cell cycle phase, thereby increasing the efficiency of homology-directed repair (HDR) in CRISPR-Cas9 genome editing (Lin et al.; Yang et al.). CANCER RESEARCH· Induces apoptosis in leukemic cells without harming T cells and/or mesenchymal stromal cells that were recovered from the same patients (Frezzato et al.).
Catalog No. 50-197-6176