Learn More
Cayman Chemical N-octanoyl-L-Homoserine lactone, Purity: ≥98%, CAS: 147852-84-4, 25 mg
SDP
Catalog No. 502887390
Encompass_Preferred
A small diffusible signaling molecule involoved in quorum sensing, controlling gene expression, and affecting cellular metabolism; applications of this molecule include infection prevention and a regulation of virulence in general and in cystic fibrosis
Catalog No. 50-288-7390 Supplier Cayman Chemical Supplier No. 1001119925 MG
A small diffusible signaling molecule involoved in quorum sensing, controlling gene expression, and affecting cellular metabolism; applications of this molecule include infection prevention and a regulation of virulence in general and in cystic fibrosis