Learn More
Cayman Chemical NoctanoylLHomoserine lactone
SDP
Catalog No. 502889299
Encompass_Preferred
A small diffusible signaling molecule involoved in quorum sensing, controlling gene expression, and affecting cellular metabolism; applications of this molecule include infection prevention and a regulation of virulence in general and in cystic fibrosis
Catalog No. 50-288-9299 Supplier Cayman Chemical Supplier No. 1001119910 MG
A small diffusible signaling molecule involoved in quorum sensing, controlling gene expression, and affecting cellular metabolism; applications of this molecule include infection prevention and a regulation of virulence in general and in cystic fibrosis