Learn More
TARGETMOL CHEMICALS INC NOD-IN-1 5MG
SDP
Catalog No. 502576910
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NOD-IN-1 (Compound 4) is a potent mixed inhibitor of nucleotide-binding oligomerization domain (NOD)-like receptors. purity: 99%
Catalog No. 50-257-6910 Supplier TARGETMOL CHEMICALS INC Supplier No. T35875MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NOD-IN-1 (Compound 4) is a potent mixed inhibitor of nucleotide-binding oligomerization domain (NOD)-like receptors. purity: 99%