Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nodakenetin | 495-32-9 | 5 MG
SDP

Catalog No. 5000421127
Encompass_Preferred

Nodakenetin is a compound with a purity of 99.66% isolated from Angelica decursiva. This white to off-white solid has demonstrated antioxidant and anti-inflammatory activities, indicating its potential utility as an antiarthritic agent and a nerve tonic.

  • Purity: 99.66%
  • Source: isolated from Angelica decursiva
  • Appearance: white to off-white solid
  • Molecular weight: 246.26
  • Formula: C14H14O4
  • CAS No.: 495-32-9
  • Biological activity: possesses antioxidant and anti-inflammatory activities
  • Potential applications: may serve as an antiarthritic agent and a nerve tonic

Catalog No. 50-004-21127 Supplier Medchemexpress LLC Supplier No. HYN22765MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nodakenetin is a compound with a purity of 99.66% isolated from Angelica decursiva. This white to off-white solid has demonstrated antioxidant and anti-inflammatory activities, indicating its potential utility as an antiarthritic agent and a nerve tonic.

  • Purity: 99.66%
  • Source: isolated from Angelica decursiva
  • Appearance: white to off-white solid
  • Molecular weight: 246.26
  • Formula: C14H14O4
  • CAS No.: 495-32-9
  • Biological activity: possesses antioxidant and anti-inflammatory activities
  • Potential applications: may serve as an antiarthritic agent and a nerve tonic

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.