Learn More
Medchemexpress LLC Nodakenin | 495-31-8 | 408.40 | 5 MG
SDP
Catalog No. 5000416999
Encompass_Preferred
Nodakenin is a significant coumarin glucoside found in the root of *Angelica decusiva*. It functions as an inhibitor of acetylcholinesterase (AChE) activity, with an IC50 of 84.7 μM. This product is intended solely for research use and is not sold to patients.
  • Nodakenin inhibits acetylcholinesterase (AChE) activity with an IC50 of 84.7 μM.
  • It is a major coumarin glucoside isolated from the root of *Angelica decusiva*.
  • Nodakenin (10 mg/kg, p.o.) reverses scopolamine-induced cognitive impairments in mice.
  • The product boasts a purity of 98.70%.
Catalog No. 50-004-16999 Supplier Medchemexpress LLC Supplier No. HYN08255MG
Nodakenin is a significant coumarin glucoside found in the root of *Angelica decusiva*. It functions as an inhibitor of acetylcholinesterase (AChE) activity, with an IC50 of 84.7 μM. This product is intended solely for research use and is not sold to patients.
  • Nodakenin inhibits acetylcholinesterase (AChE) activity with an IC50 of 84.7 μM.
  • It is a major coumarin glucoside isolated from the root of *Angelica decusiva*.
  • Nodakenin (10 mg/kg, p.o.) reverses scopolamine-induced cognitive impairments in mice.
  • The product boasts a purity of 98.70%.