Learn More
Medchemexpress LLC Nodakenin | 495-31-8 | MFCD09953919 | >98.0% | 408.40 | C20H24O9 | 10 MG
SDP
Catalog No. 5000295896
Encompass_Preferred
Nodakenin is a coumarin glucoside (C20H24O9, MW 408.40) isolated from the roots of Angelica/Peucedanum species and supplied as an analytical research standard. It is reported to inhibit acetylcholinesterase with an IC50 of 84.7 μM. Typical handling and storage recommendations include protecting from light and refrigerating; in solvent store at -80°C for long-term or -20°C for short-term storage.
  • High purity (>98% by HPLC)
  • Verified chemical identity (CAS 495-31-8)
  • Provided as a solid analytical standard suitable for research
  • Reported AChE inhibitory activity (IC50 = 84.7 μM)
  • Storage: protect from light; 4°C for solid; in solvent -80°C (6 months) or -20°C (1 month)
Catalog No. 50-002-95896 Supplier Medchemexpress LLC Supplier No. HYN082510MG
Nodakenin is a coumarin glucoside (C20H24O9, MW 408.40) isolated from the roots of Angelica/Peucedanum species and supplied as an analytical research standard. It is reported to inhibit acetylcholinesterase with an IC50 of 84.7 μM. Typical handling and storage recommendations include protecting from light and refrigerating; in solvent store at -80°C for long-term or -20°C for short-term storage.
  • High purity (>98% by HPLC)
  • Verified chemical identity (CAS 495-31-8)
  • Provided as a solid analytical standard suitable for research
  • Reported AChE inhibitory activity (IC50 = 84.7 μM)
  • Storage: protect from light; 4°C for solid; in solvent -80°C (6 months) or -20°C (1 month)