Learn More
Medchemexpress LLC HY-18639 10mg Medchemexpress, Nodinitib-1 CAS:799264-47-4 Purity:>98%
SDP
Catalog No. 502082060
Encompass_Preferred
Medchemexpress, HY-18639 10mg Nodinitib-1 CAS:799264-47-4 Nodinitib-1 (ML130;CID-1088438) is a NOD1 inhibitor with an IC50 of 0.56 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2060 Supplier Medchemexpress LLC Supplier No. HY1863910MG
Medchemexpress, HY-18639 10mg Nodinitib-1 CAS:799264-47-4 Nodinitib-1 (ML130;CID-1088438) is a NOD1 inhibitor with an IC50 of 0.56 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.