Learn More
Cayman Chemical Nodinitib-1, Purity: ≥95%, CAS: 799264-47-4, 10 mg
SDP
Catalog No. 502951098
Encompass_Preferred
Selectively inhibits NOD1-dependent activation of NF-κB and MAPK signaling (IC50 = 0.6 μM) and also inhibits NOD1-induced IL-8 production in MCF-7 cells without affecting viability
Catalog No. 50-295-1098 Supplier Cayman Chemical Supplier No. 1104010 MG
Selectively inhibits NOD1-dependent activation of NF-κB and MAPK signaling (IC50 = 0.6 μM) and also inhibits NOD1-induced IL-8 production in MCF-7 cells without affecting viability