Learn More
TARGETMOL CHEMICALS INC NODINITIB-1 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T659125MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Nodinitib-1 (CID-1088438) is a potent and selective NOD1 inhibitor with an IC50 of 0.56 uM. purity: 99%
Catalog No. 50-257-9329