Learn More
Enzo Life Sciences Nodularin (1mg). CAS: 118399-22-7
SDP
Supplier:  Enzo Life Sciences ALX350061M001
Encompass_Preferred
Inhibitor of protein phosphatase 1 (PP1) (IC50=1.8nM), protein phosphatase 2A (PP2A) (IC50=0.026nM) and to a lesser extent protein phosphatase 2B (PP2B) (IC50=8.7µm). Similar to microcystin-LR (ALX-350-012) but with increased water solubility. Genotoxic. Hepatoxic. Induces oxidative DNA damage by oxidation of purines. Induces apoptosis in HepG2 cells. Purity: ≥95% (HPLC). Appearance: Dry residue containing traces of monobasic potassium phosphate. Solubility: Soluble in methanol:water (1:1). Long Term Storage: -20°C.
Catalog No. 50-200-8712