Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nodularin Solution, 10 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111018061 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8061 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8061 Supplier MilliporeSigma Supelco Supplier No. 325391ML
Add to Cart
Add to Cart

Nodularins belong to the class of hepatotoxic cyclic pentapeptides and have their structures resembling to that of microcystins. They are produced by Nodularia spumigena cyanobacterium.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 118399-22-7
Flash Point 11°C
Linear Formula C41H60N8O10
MDL Number MFCD16659855
Synonym Cyclo[(2 S,3 S,4 E,6 E,8 S,9 S)-3-amino-9-methoxy-2,6,8-trimethyl-10-phenyl-4,6-decadienoyl-D-gamma-glutamyl-(2 Z)-2-(methylamino)-2-butenoyl-(3 S)-3-methyl-Dbeta-aspartyl-L-arginyl); Nodularin-R solution
Type Analytical Standard
Quantity 1 mL
Formula Weight 824.96
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 10 μg/mL in methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.