Learn More
Cayman Chemical Neoeriocitrin, Purity: ≥98%, CAS: 888471-94-1, 5 mg
SDP
Catalog No. 502932836
Encompass_Preferred
A flavonoid; inhibits the production of DPPH radicals by 17.2% and the formation of superoxide radicals by 48.3% in cell-free assays; decreases the rate of LDL oxidation ex vivo in isolated hamster plasma; increases proliferation of MC3T3-E1 cells and reverses the antiproliferative effect of PD 98059; increases the expression of the osteogenic differentiation markers Runx2, Type 1 collagen, and osteocalcin
Catalog No. 50-293-2836 Supplier Cayman Chemical Supplier No. 258395 MG
A flavonoid; inhibits the production of DPPH radicals by 17.2% and the formation of superoxide radicals by 48.3% in cell-free assays; decreases the rate of LDL oxidation ex vivo in isolated hamster plasma; increases proliferation of MC3T3-E1 cells and reverses the antiproliferative effect of PD 98059; increases the expression of the osteogenic differentiation markers Runx2, Type 1 collagen, and osteocalcin