Learn More
Aviva Systems Biology Corp NOG Antibody - middle region (ARP58773_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58773_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NOG. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human NOG. Alternate names: SYM1, SYNS1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat, Sheep
Catalog No. 50-286-496