Learn More
Echelon Biosciences Research Labs NOG (N-OXAL GLYCINE) 100 mg
SDP
Supplier:  Echelon Biosciences Research Labs F0020100MG
Encompass
NOG is an analogue of alpha ketoglutarate and has been found to be an inhibitor of reactions involving alpha ketoglutarate such as xanthine hydroxylase and alpha ketoglutarate dioxygenase. This item is non returnable
Catalog No. 50-720-323