Learn More
MSC Noga Dial Indicator & Base Kit, 0-100 Dial, 0.001 in, Magnetic Base
Catalog No. 5000510946
Encompass
This Dial Indicator & Base Kit features a dial indicator with a 0-100 reading and 0.001000 inch graduation, paired with a magnetic base for stable and precise measurements. It is designed for applications requiring accurate dimensional readings and instant fixation.
  • Dial indicator for precise measurements.
  • Magnetic base for secure positioning.
  • 0.001000 inch graduation for fine readings.
  • 0-100 dial reading.
  • 2-9/32 inch dial diameter.
  • Instant fixation for quick setup.
  • Bottom arm length of 56 mm.
  • Top arm length of 51 mm.
  • Includes an xlarge t-shirt.
Catalog No. 50-005-10946 Supplier MSC Supplier No. 87934295
This Dial Indicator & Base Kit features a dial indicator with a 0-100 reading and 0.001000 inch graduation, paired with a magnetic base for stable and precise measurements. It is designed for applications requiring accurate dimensional readings and instant fixation.
  • Dial indicator for precise measurements.
  • Magnetic base for secure positioning.
  • 0.001000 inch graduation for fine readings.
  • 0-100 dial reading.
  • 2-9/32 inch dial diameter.
  • Instant fixation for quick setup.
  • Bottom arm length of 56 mm.
  • Top arm length of 51 mm.
  • Includes an xlarge t-shirt.