Learn More
MSC Noga Granite Base Stand, Granite, 400 x 250 x 50 mm
Catalog No. 5000532747
Encompass
This granite base stand provides a stable and precise platform for various laboratory and industrial applications. Constructed from high-quality granite, it is designed for durability and accuracy, ensuring reliable measurements. It features fine adjustment capabilities and includes a holder for versatile use with different gauges.
  • High-quality granite construction ensures stability and precision.
  • Base dimensions are 400 mm length, 250 mm width, and 50 mm height.
  • Features fine adjustment for accurate positioning.
  • Includes a holder with a solid 3D articulated design.
  • Compatible with 6 mm, 8 mm, 3/8 in, and dovetail holding capacities.
  • Enables fast and easy setup of gauges.
Catalog No. 50-005-32747 Supplier MSC Supplier No. 20174108
This granite base stand provides a stable and precise platform for various laboratory and industrial applications. Constructed from high-quality granite, it is designed for durability and accuracy, ensuring reliable measurements. It features fine adjustment capabilities and includes a holder for versatile use with different gauges.
  • High-quality granite construction ensures stability and precision.
  • Base dimensions are 400 mm length, 250 mm width, and 50 mm height.
  • Features fine adjustment for accurate positioning.
  • Includes a holder with a solid 3D articulated design.
  • Compatible with 6 mm, 8 mm, 3/8 in, and dovetail holding capacities.
  • Enables fast and easy setup of gauges.