Learn More
MSC Noga Indicator Positioner & Holder, Articulated Arm, 72 lb Pull, 35 mm Base
Catalog No. 5000513339
Encompass
The Noga Indicator Positioner & Holder is designed for precise measurement applications. It features an articulated arm and a strong magnetic base, making it suitable for various industrial and laboratory settings.
  • Articulated arm style
  • Includes an on/off magnetic base
  • No fine adjustment feature
  • Provides a strong 72 lb magnetic pull
  • Base height of 35 mm
  • Supplied with holder and magnetic base with arm
  • Manufactured in Israel
Catalog No. 50-005-13339 Supplier MSC Supplier No. 08074379
The Noga Indicator Positioner & Holder is designed for precise measurement applications. It features an articulated arm and a strong magnetic base, making it suitable for various industrial and laboratory settings.
  • Articulated arm style
  • Includes an on/off magnetic base
  • No fine adjustment feature
  • Provides a strong 72 lb magnetic pull
  • Base height of 35 mm
  • Supplied with holder and magnetic base with arm
  • Manufactured in Israel