Learn More
MSC Noga Indicator Positioner & Holder, Fine Adjustment, Articulated Arm, 220 lb Magnetic Pull, Aluminum
Catalog No. 5000527438
Encompass
Noga Indicator Positioner & Holder is engineered with fine adjustment capabilities and an articulated arm for precise positioning. It features a magnetic base that provides a strong 220 lb pull force for secure attachment. Constructed from durable aluminum, this versatile holder also includes a V-grooved base, making it suitable for various industrial and laboratory applications requiring accurate indicator placement.
  • Articulated arm for flexible positioning.
  • Fine adjustment for precise control.
  • Magnetic base with 220 lb pull for secure attachment.
  • Constructed from aluminum for durability.
  • V-grooved base for versatile mounting.
Catalog No. 50-005-27438 Supplier MSC Supplier No. 72695455
Noga Indicator Positioner & Holder is engineered with fine adjustment capabilities and an articulated arm for precise positioning. It features a magnetic base that provides a strong 220 lb pull force for secure attachment. Constructed from durable aluminum, this versatile holder also includes a V-grooved base, making it suitable for various industrial and laboratory applications requiring accurate indicator placement.
  • Articulated arm for flexible positioning.
  • Fine adjustment for precise control.
  • Magnetic base with 220 lb pull for secure attachment.
  • Constructed from aluminum for durability.
  • V-grooved base for versatile mounting.