Learn More
MSC Noga Indicator Positioner & Holder, Fine Adjustment, Articulated Arm, Magnetic Base, 3/8 in Clamp
Catalog No. 5000513255
Encompass
This indicator positioner and holder is designed for precise positioning of indicators. It features a powerful on/off magnetic base and an articulated arm, allowing for instant fixation in various positions. This holder accommodates indicators with stem and dovetail, making it suitable for a wide range of laboratory and industrial applications.
  • Powerful on/off magnetic base.
  • Five-axis holder.
  • Holds indicators with stem and dovetail.
  • Instant fixation in any position.
  • Central locking mechanism with spring-loaded bearing.
  • Black finished arms.
  • Fine adjustment.
Catalog No. 50-005-13255 Supplier MSC Supplier No. 08074395
This indicator positioner and holder is designed for precise positioning of indicators. It features a powerful on/off magnetic base and an articulated arm, allowing for instant fixation in various positions. This holder accommodates indicators with stem and dovetail, making it suitable for a wide range of laboratory and industrial applications.
  • Powerful on/off magnetic base.
  • Five-axis holder.
  • Holds indicators with stem and dovetail.
  • Instant fixation in any position.
  • Central locking mechanism with spring-loaded bearing.
  • Black finished arms.
  • Fine adjustment.