Learn More
MSC Noga Magnetic Indicator Base, Articulated arm, 4.25 in, 70 lb pull
Catalog No. 5000529156
Encompass
This Noga Magnetic Indicator Base features an articulated arm and a powerful 70 lb magnetic pull. Designed for precision, it includes an on/off switch and a unique fine adjustment mechanism at the base, enabling faster zero-setting by allowing the hand to rest on the base. Its compact size and versatile design make it ideal for use in tight spaces and with various test indicators.
  • Unique single direction fine adjustment at the base.
  • Powerful on/off magnetic base.
  • Holds 3/8 inch and dovetail indicators.
  • Unique central locking mechanism with a spring-loaded bearing.
  • Five-axis holder provides instant fixation in any position.
  • Easy to set up into the required position.
  • Black finished arms.
Catalog No. 50-005-29156 Supplier MSC Supplier No. 09560434
This Noga Magnetic Indicator Base features an articulated arm and a powerful 70 lb magnetic pull. Designed for precision, it includes an on/off switch and a unique fine adjustment mechanism at the base, enabling faster zero-setting by allowing the hand to rest on the base. Its compact size and versatile design make it ideal for use in tight spaces and with various test indicators.
  • Unique single direction fine adjustment at the base.
  • Powerful on/off magnetic base.
  • Holds 3/8 inch and dovetail indicators.
  • Unique central locking mechanism with a spring-loaded bearing.
  • Five-axis holder provides instant fixation in any position.
  • Easy to set up into the required position.
  • Black finished arms.