Learn More
MSC Noga MG Holder, Plastic Fine Adjustment, Articulated Arm, 3/8 in Clamping Diameter
Catalog No. 5000529094
Encompass
This Noga MG holder features a unique design with plastic fine adjustment and an articulated arm, making it ideal for Electrical Discharge Machining (EDM) applications. It includes a base for secure positioning and is engineered for precise indicator placement in various industrial settings.
  • Plastic fine adjustment for precise indicator positioning.
  • Articulated arm for flexible and versatile setup.
  • Includes a base for stable mounting.
  • Clamps 3/8 in indicators and dovetail.
  • M8 external bottom thread.
  • Total arm length of 317 mm.
  • Suitable for EDM applications.
Catalog No. 50-005-29094 Supplier MSC Supplier No. 08074361
This Noga MG holder features a unique design with plastic fine adjustment and an articulated arm, making it ideal for Electrical Discharge Machining (EDM) applications. It includes a base for secure positioning and is engineered for precise indicator placement in various industrial settings.
  • Plastic fine adjustment for precise indicator positioning.
  • Articulated arm for flexible and versatile setup.
  • Includes a base for stable mounting.
  • Clamps 3/8 in indicators and dovetail.
  • M8 external bottom thread.
  • Total arm length of 317 mm.
  • Suitable for EDM applications.