Learn More
MSC Noga Replacement Knob, 1-1/2 in Long, M5x0.8 Thread, for Indicator Holders
Catalog No. 5000527938
Encompass
This replacement knob is designed for magnetic bases or indicator holders to restore smooth and precise adjustments. It provides a cost-effective solution for maintaining optimal performance of your equipment.
  • Achieves smooth adjustments for indicator holders.
  • Compatible with NF and DG series indicator holders.
  • Restores fine adjustment capabilities.
  • Offers a cost-effective replacement for lost or worn-out knobs.
  • Measures 1-1/2 in long.
  • Features an M5x0.8 thread size.
Catalog No. 50-005-27938 Supplier MSC Supplier No. 37110129
This replacement knob is designed for magnetic bases or indicator holders to restore smooth and precise adjustments. It provides a cost-effective solution for maintaining optimal performance of your equipment.
  • Achieves smooth adjustments for indicator holders.
  • Compatible with NF and DG series indicator holders.
  • Restores fine adjustment capabilities.
  • Offers a cost-effective replacement for lost or worn-out knobs.
  • Measures 1-1/2 in long.
  • Features an M5x0.8 thread size.