Learn More
Medchemexpress LLC Nogapendekin alfa h 250ug | 250ug
SDP
Catalog No. 5000322205
Encompass_Preferred
Nogapendekin alfa (his tag) a soluble protein subunit of a human interleukin (IL)-15 variant is a superagonist of IL-15 Nogapendekin alfa (his tag) promotes the proliferation and viability of immune cells Nogapendekin alfa (his tag) combines with Inbakicept (HY-P99661) at a ratio of 2 1 to form N-803 (Nogapendekin alfa inbakicept) an IL-15 cytokine antibody fusion protein N-803 reduces tumor burden by activation of NK cells and CD8 T cells[1]
Catalog No. 50-003-22205 Supplier Medchemexpress LLC Supplier No. HYP99759250UG
Nogapendekin alfa (his tag) a soluble protein subunit of a human interleukin (IL)-15 variant is a superagonist of IL-15 Nogapendekin alfa (his tag) promotes the proliferation and viability of immune cells Nogapendekin alfa (his tag) combines with Inbakicept (HY-P99661) at a ratio of 2 1 to form N-803 (Nogapendekin alfa inbakicept) an IL-15 cytokine antibody fusion protein N-803 reduces tumor burden by activation of NK cells and CD8 T cells[1]