Learn More
Medchemexpress LLC HY-P7051A 10ug Medchemexpress, Noggin (CHO-expressed), Human CAS: Purity:>98%
SDP
Catalog No. 501934297
Encompass_Preferred
Medchemexpress, HY-P7051A 10ug Noggin (CHO-expressed), Human CAS: Recombinant Human Noggin (CHO-expressed) is an antagonist of bone morphogenetic proteins (BMPs), binds with high affinity to hBMP-4, and with lower affinity to hBMP-7; Recombinant Human Noggin is essential for proper skeletal development. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4297 Supplier Medchemexpress LLC Supplier No. HYP7051A10UG
Add to Cart
Add to Cart
Medchemexpress, HY-P7051A 10ug Noggin (CHO-expressed), Human CAS: Recombinant Human Noggin (CHO-expressed) is an antagonist of bone morphogenetic proteins (BMPs), binds with high affinity to hBMP-4, and with lower affinity to hBMP-7; Recombinant Human Noggin is essential for proper skeletal development. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.