Learn More
Medchemexpress LLC HY-P7086 10ug Medchemexpress, Noggin (CHO-expressed), Mouse CAS: Purity:>98%
SDP
Catalog No. 501934326
Encompass_Preferred
Medchemexpress, HY-P7086 10ug Noggin (CHO-expressed), Mouse CAS: Recombinant Mouse Noggin (CHO-expressed) is a bone morphogenetic protein (BMP) antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4326 Supplier Medchemexpress LLC Supplier No. HYP708610UG
Medchemexpress, HY-P7086 10ug Noggin (CHO-expressed), Mouse CAS: Recombinant Mouse Noggin (CHO-expressed) is a bone morphogenetic protein (BMP) antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.