Learn More
Medchemexpress LLC Noggin Human CHO 100ug
SDP
Catalog No. 5000185062
Encompass_Preferred
Recombinant human Noggin protein, expressed in CHO cells and supplied as a lyophilized, tag-free powder for in vitro research use. Noggin binds BMP-2/4/7 to antagonize BMP signaling and is provided with high purity and formulation components to support stability and storage.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Apparent molecular weight ~27-36 kDa due to glycosylation.
  • Lyophilized formulation with trehalose for improved stability.
  • Mammalian expression (CHO) for relevant post-translational modifications.
  • Suitable for in vitro research applications; not for clinical use.
  • Multiple pack sizes available, including 100 μg.
Catalog No. 50-001-85062 Supplier Medchemexpress LLC Supplier No. HYP7051A100UG
Recombinant human Noggin protein, expressed in CHO cells and supplied as a lyophilized, tag-free powder for in vitro research use. Noggin binds BMP-2/4/7 to antagonize BMP signaling and is provided with high purity and formulation components to support stability and storage.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Apparent molecular weight ~27-36 kDa due to glycosylation.
  • Lyophilized formulation with trehalose for improved stability.
  • Mammalian expression (CHO) for relevant post-translational modifications.
  • Suitable for in vitro research applications; not for clinical use.
  • Multiple pack sizes available, including 100 μg.