Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Noggin Mouse CHO 100ug
SDP

Catalog No. 5000184858
Encompass_Preferred

Recombinant mouse Noggin protein expressed in CHO cells, supplied lyophilized and tag-free, intended for research applications requiring BMP pathway inhibition and cell-based bioactivity assays. The product is characterized by high purity, glycosylation-dependent apparent molecular weight, and reported ED50 values demonstrating potent inhibition of BMP-4-induced responses.

  • CHO-expressed, tag-free recombinant protein.
  • Lyophilized formulation with stabilizers for improved stability.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Apparent molecular weight ~28-31 kDa due to glycosylation.
  • Demonstrated bioactivity as a potent BMP inhibitor with low ED50 in cell assays.
  • Store at -20°C; reconstituted material stable at 4°C for short-term use.

Catalog No. 50-001-84858 Supplier Medchemexpress LLC Supplier No. HYP7086100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse Noggin protein expressed in CHO cells, supplied lyophilized and tag-free, intended for research applications requiring BMP pathway inhibition and cell-based bioactivity assays. The product is characterized by high purity, glycosylation-dependent apparent molecular weight, and reported ED50 values demonstrating potent inhibition of BMP-4-induced responses.

  • CHO-expressed, tag-free recombinant protein.
  • Lyophilized formulation with stabilizers for improved stability.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Apparent molecular weight ~28-31 kDa due to glycosylation.
  • Demonstrated bioactivity as a potent BMP inhibitor with low ED50 in cell assays.
  • Store at -20°C; reconstituted material stable at 4°C for short-term use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.