Learn More
Medchemexpress LLC Noggin Mouse CHO 2ug
SDP
Catalog No. 5000184200
Encompass_Preferred
Recombinant mouse Noggin (L20-C232) is a tag-free, CHO-expressed glycoprotein supplied as a lyophilized powder. It is characterized for purity, endotoxin, and specific biological activity, and is intended for cell-based assays and biochemical studies where BMP inhibition is required.
  • CHO-expressed, tag-free recombinant mouse Noggin fragment (L20-C232).
  • Glycosylated protein with apparent molecular weight ~28-31 kDa.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<0.2 EU/μg) suitable for cell assays.
  • Lyophilized formulation with stabilizers for improved storage and handling.
  • Validated biological activity with low ED50 in BMP-4 inhibition assays.
Catalog No. 50-001-84200 Supplier Medchemexpress LLC Supplier No. HYP70862UG
Recombinant mouse Noggin (L20-C232) is a tag-free, CHO-expressed glycoprotein supplied as a lyophilized powder. It is characterized for purity, endotoxin, and specific biological activity, and is intended for cell-based assays and biochemical studies where BMP inhibition is required.
  • CHO-expressed, tag-free recombinant mouse Noggin fragment (L20-C232).
  • Glycosylated protein with apparent molecular weight ~28-31 kDa.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<0.2 EU/μg) suitable for cell assays.
  • Lyophilized formulation with stabilizers for improved storage and handling.
  • Validated biological activity with low ED50 in BMP-4 inhibition assays.