Learn More
ACROBiosystems Recombinant Protein;Human Noggin Protein, Fc Tag;HEK293;1MG;NON,Noggin,NOG,SYM1,SYM-1,SYNS1,SYNS-1
Catalog No. 501657714
Encompass_Preferred
Recombinant Protein;Human Noggin Protein, Fc Tag;1MG;Human Noggin, Fc Tag (NON-H5257) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Cys 232 (Accession # NP_005441).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 49.8 kDa. The protein migrates as 58-62 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7714 Supplier ACROBiosystems Supplier No. NONH52571MG
Recombinant Protein;Human Noggin Protein, Fc Tag;1MG;Human Noggin, Fc Tag (NON-H5257) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Cys 232 (Accession # NP_005441).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 49.8 kDa. The protein migrates as 58-62 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.