Learn More
Cayman Chemical Neohesperidin, Purity: ≥98%, CAS: 13241-33-3, 100 mg
SDP
Catalog No. 502994955
Encompass_Preferred
A flavonoid with diverse biological activities; inhibits osteoclast differentiation, bone resorption, activation of NF-κB and NFAT by RANKL, and calcium oscillations in vitro; in vivo administration protects ovariectomized mice from bone loss; reduces lipid accumulation in HepG2 cells and reverses hyperlipidemia in a mouse model of diet-induced obesity; increases glucose tolerance and decreases insulin resistance while simultaneously decreasing serum triglycerides and total cholesterol and inhibiting lipid accumulation in livers in a mouse model of diabetes
Catalog No. 50-299-4955 Supplier Cayman Chemical Supplier No. 23028100 MG
A flavonoid with diverse biological activities; inhibits osteoclast differentiation, bone resorption, activation of NF-κB and NFAT by RANKL, and calcium oscillations in vitro; in vivo administration protects ovariectomized mice from bone loss; reduces lipid accumulation in HepG2 cells and reverses hyperlipidemia in a mouse model of diet-induced obesity; increases glucose tolerance and decreases insulin resistance while simultaneously decreasing serum triglycerides and total cholesterol and inhibiting lipid accumulation in livers in a mouse model of diabetes