Learn More
Cayman Chemical Neohesperidin dihydrochalcone, Purity: ≥98%, CAS: 20702-77-6, 25 g
SDP
Catalog No. 502939508
Encompass_Preferred
A semisynthetic glycoside chalcone, an artificial sweetener, and a derivative of neohesperidin; scavenges 59.9, 66.7, 6.3, and 20% of DPPH, ABTS, superoxide, and hydroxyl radicals, respectively, in cell-free assays at 50 μM; inhibits decreases in hepatic GSH levels and increases in serum AST and ALT activity and hepatic levels of ROS, TBARS, NF-κB, IL-6, IL-1β, and TNF-α in a mouse model of paraquat-induced acute liver at 200 mg/kg per day
Catalog No. 50-293-9508 Supplier Cayman Chemical Supplier No. 2851025 G
A semisynthetic glycoside chalcone, an artificial sweetener, and a derivative of neohesperidin; scavenges 59.9, 66.7, 6.3, and 20% of DPPH, ABTS, superoxide, and hydroxyl radicals, respectively, in cell-free assays at 50 μM; inhibits decreases in hepatic GSH levels and increases in serum AST and ALT activity and hepatic levels of ROS, TBARS, NF-κB, IL-6, IL-1β, and TNF-α in a mouse model of paraquat-induced acute liver at 200 mg/kg per day