Learn More
Perkin Elmer US LLC Noise Enclosure for Polyscience 6000 Chiller
Catalog No. 502104122
Encompass_Preferred
Performance: average sound level decrease is - 15 dB(A) (equivalent to a 75% reduction in noise perception) Environment friendly noise adsorbing material, oil and water resistant. Heat exchange controlled by six internal silent fans Worldwide electrical compatibility-enclosures are delivered with European, US, Japan, and UK plugs Easy access to front monitor and switch (the transparent window can be opened). Easy water level check (window on the back panel). Includes Overheating temperature alarm The noise reduction is in conformity with the European directive related to low voltage equipment(2006/95/EC) and electromagnetic compatibility (2004/108/EC). Power supply Csa, Ce and Ccc
Catalog No. 50-210-4122 Supplier Perkin Elmer US LLC Supplier No. N0777920
Add to Cart
Add to Cart
Request a Quote
Performance: average sound level decrease is - 15 dB(A) (equivalent to a 75% reduction in noise perception) Environment friendly noise adsorbing material, oil and water resistant. Heat exchange controlled by six internal silent fans Worldwide electrical compatibility-enclosures are delivered with European, US, Japan, and UK plugs Easy access to front monitor and switch (the transparent window can be opened). Easy water level check (window on the back panel). Includes Overheating temperature alarm The noise reduction is in conformity with the European directive related to low voltage equipment(2006/95/EC) and electromagnetic compatibility (2004/108/EC). Power supply Csa, Ce and Ccc