Learn More
Chemglass Life Sciences Cap w/hole, SVL, Size 15, Fits Tubing 5.60-6.40mm OD
SDP
Catalog No. CG18901
Encompass_Preferred
Cap w/hole, SVL, Size 15, Fits Tubing 5.60-6.40mm OD. Open top SVL screw cap supplied complete with PTFE sealing ring to accept the tubing size as listed.
Catalog No. CG18901 Supplier Chemglass Life Sciences Supplier No. CG18901
Cap w/hole, SVL, Size 15, Fits Tubing 5.60-6.40mm OD. Open top SVL screw cap supplied complete with PTFE sealing ring to accept the tubing size as listed.