Learn More
Chemglass Life Sciences Solid Cap, SVL, Size 15
SDP
Catalog No. CG18801
Encompass_Preferred
Solid Cap, SVL, Size 15. Solid SVL cap for use with CG-187 screw threads. Caps come complete with PTFE faced liner. Cap and liner are autoclavable to 121C.
Catalog No. CG18801 Supplier Chemglass Life Sciences Supplier No. CG18801
Solid Cap, SVL, Size 15. Solid SVL cap for use with CG-187 screw threads. Caps come complete with PTFE faced liner. Cap and liner are autoclavable to 121C.