Learn More
Aviva Systems Biology Corp NOL5A Antibody - middle region (ARP40645_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40645_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NOL5A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NOL5A. Alternate names: RP4-686C3.1, NOP56, NOL5A, SCA36ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57568