Learn More
Aviva Systems Biology Corp NOL6 Antibody - C-terminal region (ARP40996_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40996_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NOL6. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human NOL6. Alternate names: NRAP, UTP22, bA311H10.1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57691