Learn More
Chemglass Life Sciences Solid Cap, PTFE Seal, GL-18 Thread
SDP
Catalog No. CG19602
Encompass_Preferred
Solid Cap, PTFE Seal, GL-18 Thread. Cap is made from PBT (thermoplastic polyester) and will withstand temperatures from -45 to 200º C. PTFE seal is also available separately.
Catalog No. CG19602 Supplier Chemglass Life Sciences Supplier No. CG19602
Solid Cap, PTFE Seal, GL-18 Thread. Cap is made from PBT (thermoplastic polyester) and will withstand temperatures from -45 to 200º C. PTFE seal is also available separately.