Learn More
Medchemexpress LLC HY-108474 10mg Medchemexpress, Nolatrexed dihydrochloride CAS:152946-68-4 Purity:>98%
SDP
Catalog No. 501968557
Encompass_Preferred
Medchemexpress, HY-108474 10mg Nolatrexed diHCl CAS:152946-68-4 Nolatrexed dihydrochloride (AG 337) is a non-competitive lipophilic inhibitor of thymidylate synthase, interacts at the folate cofactor binding site of the enzyme, with a Ki of 11 nM for human thymidylate synthase. Nolatrexed dihydrochloride (AG 337) has anti-cancer activity, induces cell cycle arrest in S phase of cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8557 Supplier Medchemexpress LLC Supplier No. HY10847410MG
Medchemexpress, HY-108474 10mg Nolatrexed diHCl CAS:152946-68-4 Nolatrexed dihydrochloride (AG 337) is a non-competitive lipophilic inhibitor of thymidylate synthase, interacts at the folate cofactor binding site of the enzyme, with a Ki of 11 nM for human thymidylate synthase. Nolatrexed dihydrochloride (AG 337) has anti-cancer activity, induces cell cycle arrest in S phase of cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.