Learn More
Medchemexpress LLC Nolatrexed dihydrochloride | 152946-68-4 | 98.5% | 357.26 | 50 MG
SDP
Catalog No. 5000347171
Encompass_Preferred
Nolatrexed dihydrochloride (AG 337) is an orally active and non-competitive lipophilic inhibitor of thymidylate synthase with a Ki of 11 nM for human thymidylate synthase. It interacts at the folate cofactor binding site of the enzyme. Nolatrexed dihydrochloride increases sulfotransferases and induces cell cycle arrest in the S phase of cancer cells. It also exhibits anticancer activity against cervical cancer.
  • Orally active and non-competitive lipophilic inhibitor of thymidylate synthase.
  • Interacts at the folate cofactor binding site of the enzyme.
  • Increases sulfotransferases.
  • Induces cell cycle arrest in the S phase of cancer cells.
  • Exhibits anticancer activity against cervical cancer.
Catalog No. 50-003-47171 Supplier Medchemexpress LLC Supplier No. HY10847450MG
Nolatrexed dihydrochloride (AG 337) is an orally active and non-competitive lipophilic inhibitor of thymidylate synthase with a Ki of 11 nM for human thymidylate synthase. It interacts at the folate cofactor binding site of the enzyme. Nolatrexed dihydrochloride increases sulfotransferases and induces cell cycle arrest in the S phase of cancer cells. It also exhibits anticancer activity against cervical cancer.
  • Orally active and non-competitive lipophilic inhibitor of thymidylate synthase.
  • Interacts at the folate cofactor binding site of the enzyme.
  • Increases sulfotransferases.
  • Induces cell cycle arrest in the S phase of cancer cells.
  • Exhibits anticancer activity against cervical cancer.