Learn More
Cayman Chemical Nolatrexed (hydrochloride), Purity: ≥98%, CAS: 152946-68-4, 50 mg
SDP
Catalog No. 502937235
Encompass_Preferred
A thymidylate synthase inhibitor (Ki = 11 nM); inhibits cell growth in a panel of human and murine cancer cell lines (IC50s = 0.39-6.6 μM); inhibits the growth of L1210 cells, an effect that is decreased by greater than 50-fold by the addition of exogenous thymidine; induces cell cycle arrest at the S phase in L1210 cells at 0.7 and 3.5 μM; increases life span in a thymidine kinase-deficient L5178Y/TK- murine lymphoma model at 25-200 mg/kg
Catalog No. 50-293-7235 Supplier Cayman Chemical Supplier No. 2924750 MG
A thymidylate synthase inhibitor (Ki = 11 nM); inhibits cell growth in a panel of human and murine cancer cell lines (IC50s = 0.39-6.6 μM); inhibits the growth of L1210 cells, an effect that is decreased by greater than 50-fold by the addition of exogenous thymidine; induces cell cycle arrest at the S phase in L1210 cells at 0.7 and 3.5 μM; increases life span in a thymidine kinase-deficient L5178Y/TK- murine lymphoma model at 25-200 mg/kg